2-(4-Pyridinyl)-4-thiazolecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B20457-03 B20457-06
CAAAB20457-03EA 63.36 CAD
CAAAB20457-03 CAAAB20457-06
2-(4-Pyridinyl)-4-thiazolecarboxylic acid 97%
2-(4-Pyridinyl)-4-thiazolecarboxylic acid

Purity: 97% CAS No.: 21278-86-4 Molecular Formula: C9H6N2O2S Formula Weight: 206.22 Melting Point: 300° dec. MDL No.: MFCD00171745

Formula: C₉H₆N₂O₂S
MW: 206.22 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00171745
CAS Number: 21278-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR