4-Methyl-1,2,3-thiadiazole-5-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20449-06
B20449-14
CAAAB20449-06EA
186.44
CAD
CAAAB20449-06
CAAAB20449-14
CAAAB20449-22
4-Methyl-1,2,3-thiadiazole-5-carboxylic acid 98%
4-Methyl-1,2,3-thiadiazole-5-carboxylic acid
Purity: 98%
CAS No.: 18212-21-0
Molecular Formula: C4H4N2O2S
Formula Weight: 144.15
Melting Point: 184-186°
MDL No.: MFCD00173830
Formula:
C₄H₄N₂O₂S MW: 144.15 g/mol Melting Pt: 184…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173830 CAS Number: 18212-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...