3-Methyl-5-hexen-3-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

B20447-03 B20447-06 B20447-14
CAAAB20447-03EA 27.02 CAD
CAAAB20447-03 CAAAB20447-06 CAAAB20447-14 CAAAB20447-22
3-Methyl-5-hexen-3-ol 98%
3-Methyl-5-hexen-3-ol

Purity: 98% CAS No.: 1569-44-4 Molecular Formula: C7H14O Formula Weight: 114.19 Flash Point: 44°(111°F) Boiling Point: 138-139° UN No.: 1987 MDL No.: MFCD00021826

Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 138…139 °C
Density: 0.84
Flash Pt: 44 °C (111 °F)
Storage Temperature: Ambient
MDL Number: MFCD00021826
CAS Number: 1569-44-4
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR