5-Mercapto-1-methyl-1H-tetrazole 98%
Supplier: Thermo Scientific Chemicals
B20443-06
B20443-14
CAAAB20443-06EA
56.56
CAD
CAAAB20443-06
CAAAB20443-14
5-Mercapto-1-methyl-1H-tetrazole 98%
5-Mercapto-1-methyl-1H-tetrazole
Purity: 98%
CAS No.: 13183-79-4
Molecular Formula: C2H4N4S
Formula Weight: 116.14
Melting Point: 125-128°
MDL No.: MFCD00037317
Formula:
C₂H₄N₄S Melting Pt: 125…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037317 CAS Number: 13183-79-4 EINECS: 236-132-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...