2-(2-Naphthyl)indole 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(Naphthalen-2-yl)-1H-indole

B20434-03 B20434-06
CAAAB20434-03EA 119.92 CAD
CAAAB20434-03 CAAAB20434-06
2-(2-Naphthyl)indole 98%
2-(2-Naphthyl)indole

Purity: 98% CAS No.: 23746-81-8 Molecular Formula: C18H13N Formula Weight: 243.31 MDL No.: MFCD00124144

Formula: C₁₈H₁₃N
MW: 243.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00124144
CAS Number: 23746-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR