Nicotinamidine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20406-03
B20406-06
CAAAB20406-03EA
39.82
CAD
CAAAB20406-03
CAAAB20406-06
CAAAB20406-14
Nicotinamidine hydrochloride 97%
Nicotinamidine hydrochloride
Purity: 97%
CAS No.: 7356-60-7
Molecular Formula: C6H7N3.HCl
Formula Weight: 157.60
Melting Point: 187-189°
MDL No.: MFCD00054326
Formula:
C₆H₈ClN₃ Melting Pt: 187…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00054326 CAS Number: 7356-60-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...