2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydrazinyl-4,5-dihydro-1H-imidazole hydrobromide, 2-Hydrazino-2-imidazoline Hydrobromide, 2-Hydrazino-4,5-dihydroimidazole Hydrobromide
B20386-09
B20386-88
CAAAB20386-09EA
350.76
CAD
CAAAB20386-09
CAAAB20386-88
2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide 98%
2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide
Purity: 98%
CAS No.: 55959-84-7
Molecular Formula: C3H8N4.HBr
Formula Weight: 181.04
Melting Point: 181-185°
MDL No.: MFCD00012694
Formula:
C₃H₉BrN₄ MW: 181.04 g/mol Melting Pt: 181…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012694 CAS Number: 55959-84-7 EINECS: 259-921-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...