Sodium 1-hexadecanesulfonate 97%

Supplier: Thermo Scientific Chemicals

B20382-22 B20382-06 B20382-14
CAAAAB20382-22EA 1052.28 CAD
CAAAAB20382-22 CAAAB20382-06 CAAAB20382-14
Sodium 1-hexadecanesulfonate 97%
Sodium 1-hexadecanesulphonate

Purity: 97% CAS No.: 15015-81-3 Molecular Formula: C16H33SO3Na Formula Weight: 328.49 Beilstein Reference: 3920654 MDL No.: MFCD00007528

Formula: C₁₆H₃₃NaSO₃
Storage Temperature: Ambient
MDL Number: MFCD00007528
CAS Number: 15015-81-3
EINECS: 239-100-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR