3-Hydroxy-1-adamantane carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B20377-03 B20377-06
CAAAB20377-03EA 119.13 CAD
CAAAB20377-03 CAAAB20377-06
3-Hydroxy-1-adamantane carboxylic acid 97%
3-Hydroxy-1-adamantane carboxylic acid

Purity: 97% CAS No.: 42711-75-1 Molecular Formula: C11H16O3 Formula Weight: 196.25 Melting Point: 203-207° MDL No.: MFCD00193929

Formula: C₁₁H₁₆O₃
Melting Pt: 203…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00193929
CAS Number: 42711-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR