2-Methyl-3-phenyl-1-propene 99%

Supplier: Thermo Scientific Chemicals

B20370-06 B20370-14
CAAAB20370-06EA 156.26 CAD
CAAAB20370-06 CAAAB20370-14
2-Methyl-3-phenyl-1-propene 99%
2-Methyl-3-phenyl-1-propene

Purity: 99% CAS No.: 3290-53-7 Molecular Formula: C10H12 Formula Weight: 132.21 MDL No.: MFCD00039815

Formula: C₁₀H₁₂
Storage Temperature: Ambient
MDL Number: MFCD00039815
CAS Number: 3290-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR