Potassium-1-carboxyvinyl hydrogenphosphate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Phosphoenolpyruvate potassium salt, Potassium 1-carboxyvinyl hydrogenphosphate, Phosphoenolpyruvic acid monopotassium salt, Phosphoenolpyruvic acid potassium

B20358-06 B20358-14 B20358-22
CAAAB20358-06EA 331.8 CAD
CAAAB20358-06 CAAAB20358-14 CAAAB20358-22
Potassium-1-carboxyvinyl hydrogenphosphate 99%
Potassium-1-carboxyvinyl hydrogenphosphate

Purity: 99% CAS No.: 4265-07-0 Molecular Formula: C3H4KO6P Formula Weight: 206.14 Storage Temperature: -30°C to -10°C Melting Point: ca 170° dec. Beilstein Reference: 4603446 MDL No.: MFCD00044476

Formula: C₃H₄KO₆P
MW: 206.131 g/mol
Melting Pt: ∼ 170 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00044476
CAS Number: 4265-07-0
EINECS: 224-247-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR