Sodium 1-undecanesulfonate, (max. 2% H₂O) 98+% (dry weight)

Supplier: Thermo Scientific Chemicals

B20332-22 B20332-14
CAAAAB20332-22EA 1036.48 CAD
CAAAAB20332-22 CAAAB20332-03 CAAAB20332-06 CAAAB20332-14
Sodium 1-undecanesulfonate, (max. 2% H₂O) 98+% (dry weight)
Sodium 1-undecanesulphonate

Purity: 98+% CAS No.: 5838-34-6 Molecular Formula: CH3(CH2)10SO3Na Formula Weight: 258.36 Melting Point: 300° Beilstein Reference: 3751377

Formula: C₁₂H₂₅NaO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00053627
CAS Number: 5838-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR