1-tert-Butyl-3-ethylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

B20325-06 B20325-14
CAAAB20325-06EA 111.71 CAD
CAAAB20325-06 CAAAB20325-14
1-tert-Butyl-3-ethylbenzene 98%
1-tert-Butyl-3-ethylbenzene

Purity: 98% CAS No.: 14411-56-4 Molecular Formula: C12H18 Formula Weight: 162.27 Odor: Mild Boiling Point: 205° MDL No.: MFCD00060845

Formula: C₁₂H₁₈
Boiling Pt: 205 °C
Density: 0,86
Storage Temperature: Ambient
MDL Number: MFCD00060845
CAS Number: 14411-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR