4-(Trifluoromethylthio)benzaldehyde 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20313-03
B20313-06
CAAAB20313-03EA
82
CAD
CAAAB20313-03
CAAAB20313-06
4-(Trifluoromethylthio)benzaldehyde 90+%
4-(Trifluoromethylthio)benzaldehyde
Purity: 90+%
CAS No.: 4021-50-5
Molecular Formula: C8H5F3OS
Formula Weight: 206.19
Flash Point: 96°(204°F)
Refractive Index: 1.5110
Odor: Stench
Melting Point: 29-31°
Boiling Point: 219°
Beilstein Reference: 2328411
MDL No.: MFCD00129191
Formula:
C₈H₅F₃OS Boiling Pt: 219 °C Melting Pt: 29…31 °C Density: 1.310 Flash Pt: 96 °C(204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00129191 CAS Number: 4021-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...