1-(4-Chlorophenyl)biguanide hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

B20311-03 B20311-06
CAAAB20311-03EA 47.87 CAD
CAAAB20311-03 CAAAB20311-06
1-(4-Chlorophenyl)biguanide hydrochloride 97%
1-(4-Chlorophenyl)biguanide hydrochloride

Purity: 97% CAS No.: 4022-81-5 Molecular Formula: C8H10ClN5.HCl Formula Weight: 248.12 Melting Point: 247-250° Beilstein Reference: 3732533

Formula: C₈H₁₁Cl₂N₅
Melting Pt: 247…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00053020
CAS Number: 4022-81-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR