Sodium 4-bromobenzenesulfinate dihydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

B20281-09 B20281-88
CAAAB20281-09EA 298.62 CAD
CAAAB20281-09 CAAAB20281-18 CAAAB20281-88
Sodium 4-bromobenzenesulfinate dihydrate 97%
Sodium 4-bromobenzenesulphinate dihydrate

Purity: 97%% CAS No.: 175278-64-5 Molecular Formula: C6H4BrNaO2S.2H2O Formula Weight: 279.09 (243.06anhy) Melting Point: 280° MDL No.: MFCD00210139

Formula: C₆H₄BrNaO₂S·2H₂O
Melting Pt: >280 °C
Storage Temperature: Ambient
MDL Number: MFCD00210139
CAS Number: 175278-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR