DL-Asparagine monohydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: DL-Aspartic acid 4-amide monohydrate, H-DL-Asn-OH monohydrate, (±)-2-Aminosuccinic acid 4-amide monohydrate, DL-Asparagine hydrate, H-DL-Asn-OH·H₂O, DL-2-Aminosuccinamic acid monohydrate, 2,4-Diamino-4-oxobutanoic acid monohydrate

B20273-22 B20273-36
CAAAB20273-22EA 116.92 CAD
CAAAB20273-22 CAAAB20273-36
DL-Asparagine monohydrate 98%
DL-Asparagine monohydrate

Purity: 98% CAS No.: 3130-87-8 Molecular Formula: C4H8N2O3.H2O Formula Weight: 150.14 (132.12anhy) Melting Point: ca 220° dec. Beilstein Reference: 6234992 MDL No.: MFCD00151039

Formula: C₄H₈N₂O₃·H₂O
MW: 150.13 g/mol
Melting Pt: ∼ 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151039
CAS Number: 69833-18-7
EINECS: 221-521-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR