Methyl-4-bromo-3-hydroxythiophene-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

B20249-09 B20249-88
CAAAB20249-09EA 415.54 CAD
CAAAB20249-09 CAAAB20249-88
Methyl-4-bromo-3-hydroxythiophene-2-carboxylate 97%
Methyl 4-bromo-3-hydroxythiophene-2-carboxylate

Purity: 97% CAS No.: 95201-93-7 Molecular Formula: C6H5BrO3S Formula Weight: 237.08 Melting Point: 79-80° MDL No.: MFCD00052081

Formula: C₆H₅BrO₃S
Melting Pt: 79…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00052081
CAS Number: 95201-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR