1-(3,5-Bis(trifluoromethyl)phenyl)thiourea 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-[3,5-Bis(trifluoromethyl)phenyl]thiourea
B20245-03
B20245-06
CAAAB20245-03EA
65.57
CAD
CAAAB20245-03
CAAAB20245-06
1-(3,5-Bis(trifluoromethyl)phenyl)thiourea 98+%
1-(3,5-Bis(trifluoromethyl)phenyl)thiourea
Purity: 98+%
CAS No.: 175277-17-5
Molecular Formula: C9H6F6N2S
Formula Weight: 288.22
Melting Point: 168-172°
UN No.: 2811
MDL No.: MFCD00041172
Formula:
C₉H₆F₆N₂S MW: 288.22 Melting Pt: 168…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041172 CAS Number: 175277-17-5 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...