3,4-Dimethyl-2-hexanol erythro + threo 98%

Supplier: Thermo Scientific Chemicals
Warning

B20243-03 B20243-14
CAAAB20243-03EA 51.35 CAD
CAAAB20243-03 CAAAB20243-06 CAAAB20243-14
3,4-Dimethyl-2-hexanol erythro + threo 98%
3,4-Dimethyl-2-hexanol

Purity: 98% CAS No.: 31350-88-6 Molecular Formula: C8H18O Formula Weight: 130.23 MDL No.: MFCD00021887

Formula: C₈H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD00021887
CAS Number: 31350-88-6
EINECS: 631-563-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR