2-Methoxyphenyl acetate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-Acetoxy-2-methoxybenzene, Acetic acid 2-methoxyphenyl ester, 2-Acetoxyanisole, Acetic acid 2-guaiacol ester, 2-Guaiacol acetate, O-Acetylguaiacol
B20242-14
B20242-22
CAAAB20242-14EA
52.61
CAD
CAAAB20242-14
CAAAB20242-22
2-Methoxyphenyl acetate 98%
2-Methoxyphenyl acetate
Purity: 98%
CAS No.: 15212-03-0
Molecular Formula: C9H10O3
Formula Weight: 166.18
MDL No.: MFCD00017221
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00017221 CAS Number: 15212-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...