Methyl (3-methoxyphenyl)acetate 97%
Supplier: Thermo Scientific Chemicals
B20237-09
B20237-88
CAAAB20237-09EA
316
CAD
CAAAB20237-09
CAAAB20237-88
Methyl (3-methoxyphenyl)acetate 97%
Methyl (3-methoxyphenyl)acetate
Purity: 97%
CAS No.: 18927-05-4
Molecular Formula: C10H12O3
Formula Weight: 180.23
Boiling Point: 86-88°/0.5mm
MDL No.: MFCD00017205
Formula:
C₁₀H₁₂O₃ Boiling Pt: 86…88 °C/0.5 mm Storage Temperature: Ambient |
MDL Number:
MFCD00017205 CAS Number: 18927-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...