Methyl-3-(methylamino)but-2-enoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20199-06
B20199-14
CAAAB20199-03EA
97.85
CAD
CAAAB20199-03
CAAAB20199-06
CAAAB20199-14
Methyl-3-(methylamino)but-2-enoate 97%
Methyl-3-(methylamino)but-2-enoate
Purity: 97%
CAS No.: 13412-12-9
Molecular Formula: CH3C(NHCH3)=CHCO2CH3
Formula Weight: 129.16
Melting Point: 60-63°
MDL No.: MFCD00027383
Formula:
C₆H₁₁NO₂ Melting Pt: 60…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00027383 CAS Number: 13412-12-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...