2-Hydroxy-6-methylnicotinonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 3-Cyano-6-methyl-2-pyridone
B20188-09
B20188-18
B20188-30
CAAAB20188-09EA
100.49
CAD
CAAAB20188-09
CAAAB20188-18
CAAAB20188-30
2-Hydroxy-6-methylnicotinonitrile 98%
2-Hydroxy-6-methylnicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Purity: 98%
CAS No.: 4241-27-4
Molecular Formula: C7H6N2O
Formula Weight: 134.14
Melting Point: ca 298° dec.
Beilstein Reference: 120412
Formula:
C₇H₆N₂O MW: 134.14 g/mol Melting Pt: ∼ 298 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006013 CAS Number: 4241-27-4 EINECS: 224-202-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...