Cyclopropanecarboxamidine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20185-14
B20185-06
CAAAAB20185-14EA
820.02
CAD
CAAAAB20185-14
CAAAB20185-03
CAAAB20185-06
Cyclopropanecarboxamidine hydrochloride 97%
Cyclopropanecarboxamidine hydrochloride
Purity: 97%
CAS No.: 57297-29-7
Molecular Formula: C4H8N2.HCl
Formula Weight: 120.59
Melting Point: 122-123°
Beilstein Reference: 4507255
MDL No.: MFCD00053010
Formula:
C₄H₈N₂ Melting Pt: 122…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053010 CAS Number: 57297-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...