Ethyl diacetoacetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diacetoacetic Acid Ethyl Ester, Ethyl 2-Acetyl-3-oxobutyrate, Ethyl 2-acetyl-3-oxobutanoate
B20149-09
B20149-18
CAAAAB20149-30EA
1609.07
CAD
CAAAAB20149-30
CAAAB20149-09
CAAAB20149-18
Ethyl diacetoacetate 97%
Ethyl diacetoacetate
Purity: 97%
CAS No.: 603-69-0
Molecular Formula: C8H12O4
Formula Weight: 172.18
Flash Point: 85°(185°F)
Refractive Index: 1.4720
Boiling Point: 104-105°/17mm
Beilstein Reference: 775014
MDL No.: MFCD00009864
Formula:
C₈H₁₂O₄ MW: 172.18 g/mol Boiling Pt: 104…105 °C/17 mm Density: 1.104 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009864 CAS Number: 603-69-0 EINECS: 210-053-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...