5-Methyl-3-phenyl-1,2,4-oxadiazole 97%
Supplier: Thermo Scientific Chemicals
B20145-03
B20145-06
CAAAB20145-03EA
118.97
CAD
CAAAB20145-03
CAAAB20145-06
5-Methyl-3-phenyl-1,2,4-oxadiazole 97%
5-Methyl-3-phenyl-1,2,4-oxadiazole
Purity: 97%
CAS No.: 1198-98-7
Molecular Formula: C9H8N2O
Formula Weight: 223.28
Melting Point: 33-36°
MDL No.: MFCD00085133
Formula:
C₉H₈N₂O Melting Pt: 33…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD00085133 CAS Number: 1198-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...