3-Amino-4-bromo-5-methylisoxazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20138-03
B20138-06
CAAAB20138-03EA
126.08
CAD
CAAAB20138-03
CAAAB20138-06
CAAAB20138-14
3-Amino-4-bromo-5-methylisoxazole 97%
3-Amino-4-bromo-5-methylisoxazole
Purity: 97%
CAS No.: 5819-40-9
Molecular Formula: C4H5BrN2O
Formula Weight: 177.02
Melting Point: 73-75°
MDL No.: MFCD00052553
Formula:
C₄H₅BrN₂O Melting Pt: 73…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052553 CAS Number: 5819-40-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...