6-Iodo-1,4-benzodioxane 95% remainder mainly 5-isomer

Supplier: Thermo Scientific Chemicals

Synonyms: 6-Iodo-2,3-dihydro-1,4-benzodioxine, 1,2-Ethylenedioxy-4-iodobenzene, 6-Iodo-2,3-dihydrobenzo[b][1,4]dioxine

B20127-06 B20127-14
CAAAB20127-06EA 83.9 CAD
CAAAB20127-06 CAAAB20127-14
6-Iodo-1,4-benzodioxane 95% remainder mainly 5-isomer
6-Iodo-1,4-benzodioxane

Purity: 95% CAS No.: 57744-67-9 Molecular Formula: C8H7IO2 Formula Weight: 262.05 Boiling Point: 100-102°/0.25mm MDL No.: MFCD00221460

Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 100…102 °C/0.25 mm
Storage Temperature: Ambient
MDL Number: MFCD00221460
CAS Number: 57744-67-9
EINECS: 257-817-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR