2,3-Diphenylquinoxaline 98+%
Supplier: Thermo Scientific Chemicals
B20100-22
B20100-06
B20100-14
CAAAAB20100-22EA
406.06
CAD
CAAAAB20100-22
CAAAB20100-06
CAAAB20100-14
2,3-Diphenylquinoxaline 98+%
2,3-Diphenylquinoxaline
Purity: 98+%
CAS No.: 1684-14-6
Molecular Formula: C20H14N2
Formula Weight: 282.35
Melting Point: 124-128°
Beilstein Reference: 209272
Formula:
C₂₀H₁₄N₂ Melting Pt: 124…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014659 CAS Number: 1684-14-6 EINECS: 216-865-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...