5-Amino-3-tert-butyl-1-methylpyrazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20095-03
B20095-06
B20095-14
CAAAB20095-03EA
111.71
CAD
CAAAB20095-03
CAAAB20095-06
CAAAB20095-14
5-Amino-3-tert-butyl-1-methylpyrazole 98%
5-Amino-3-tert-butyl-1-methylpyrazole
Purity: 98%
CAS No.: 118430-73-2
Molecular Formula: C8H15N3
Formula Weight: 153.22
Melting Point: 155-158°
MDL No.: MFCD00068002
Formula:
C₈H₁₅N₃ Melting Pt: 155…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068002 CAS Number: 118430-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...