1,3-Propanedisulfonic acid disodium salt 99% (dry weight)

Supplier: Thermo Scientific Chemicals
Danger

B20092-06 B20092-14
CAAAB20092-06EA 73.63 CAD
CAAAB20092-06 CAAAB20092-14
1,3-Propanedisulfonic acid disodium salt 99% (dry weight)
1,3-Propanedisulphonic acid disodium salt

Purity: 99% CAS No.: 36589-58-9 Molecular Formula: NaO3S(CH2)3SO3Na Formula Weight: 248.19 Melting Point: 300° Beilstein Reference: 3767479

Formula: C₃H₆Na₂O₆S₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00191483
CAS Number: 36589-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR