2-Amino-4-methylthiophene-3-carboxamide 98%
Supplier: Thermo Scientific Chemicals
B20043-03
B20043-06
CAAAB20043-03EA
95.12
CAD
CAAAB20043-03
CAAAB20043-06
CAAAB20043-14
2-Amino-4-methylthiophene-3-carboxamide 98%
2-Amino-4-methylthiophene-3-carboxamide
Purity: 98%
CAS No.: 4651-97-2
Molecular Formula: C6H8N2OS
Formula Weight: 156.20
Melting Point: 173-175°
MDL No.: MFCD00052594
Formula:
C₆H₈N₂OS MW: 156.20 g/mol Melting Pt: 173…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052594 CAS Number: 4651-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...