Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20028-03
B20028-06
CAAAB20028-03EA
142.04
CAD
CAAAB20028-03
CAAAB20028-06
Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride 97%
Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride
Purity: 97%
CAS No.: 121071-71-4
Molecular Formula: C8H9NO4S.HCl
Formula Weight: 251.69
Melting Point: 166-169°
MDL No.: MFCD00205189
Formula:
C₈H₁₀ClNO₄S Melting Pt: 166…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00205189 CAS Number: 121071-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...