4-Hydrazinobenzenesulfonamide hydrochloride 97%
Supplier: Thermo Scientific Chemicals
B20015-09
B20015-18
B20015-88
CAAAB20015-09EA
77.1
CAD
CAAAB20015-09
CAAAB20015-18
CAAAB20015-88
4-Hydrazinobenzenesulfonamide hydrochloride 97%
4-Hydrazinobenzenesulfonamide hydrochloride
Purity: 97%
CAS No.: 17852-52-7
Molecular Formula: C9H9N3O2S.HCl
Formula Weight: 223.68
Melting Point: 217-219° dec.
MDL No.: MFCD00052262
Formula:
C₆H₉N₃O₂S Melting Pt: 217…219 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00052262 CAS Number: 17852-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...