2-Chloroethanesulfonic acid sodium salt hydrate, (max. 10% H₂O) 98+% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium-2-chloroethanesulphonate hydrate, Sodium-2-chloroethanesulfonate hydrate

A19995-06 A19995-14 A19995-22
CAAAA19995-06EA 56.72 CAD
CAAAA19995-06 CAAAA19995-14 CAAAA19995-22
2-Chloroethanesulfonic acid sodium salt hydrate, (max. 10% H₂O) 98+% (dry weight)
2-Chloroethanesulfonic acid sodium salt hydrate

Purity: 98+% CAS No.: 15484-44-3 Molecular Formula: Cl(CH2)2SO3Na.xH2O Formula Weight: 166.56(anhy) Melting Point: ca 292° dec. Beilstein Reference: 5649202

Formula: C₂H₅ClO₃S·H₂O
MW: 184.57 g/mol
Melting Pt: ∼ 292 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007531
CAS Number: 15484-44-3
EINECS: 239-508-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR