Morpholine-4-carboxamidine hydrobromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19985-06
A19985-14
A19985-22
CAAAA19985-06EA
173.8
CAD
CAAAA19985-06
CAAAA19985-14
CAAAA19985-22
Morpholine-4-carboxamidine hydrobromide 98%
Morpholine-4-carboxamidine hydrobromide
Purity: 98%
CAS No.: 157415-17-3
Molecular Formula: C5H11N3O.HBr
Formula Weight: 210.08
Melting Point: ca 155-165°
Beilstein Reference: 7389038
MDL No.: MFCD00051994
Formula:
C₅H₁₂BrN₃O Melting Pt: ca 155…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051994 CAS Number: 157415-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...