Cyclopentyl phenyl ketone 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Cyclopentyl(phenyl)methanone, Benzoylcyclopentane

A19976-03 A19976-06
CAAAA19976-03EA 51.98 CAD
CAAAA19976-03 CAAAA19976-06
Cyclopentyl phenyl ketone 96%
Cyclopentyl phenyl ketone

Purity: 96% CAS No.: 5422-88-8 Molecular Formula: C12H14O Formula Weight: 174.24 Flash Point: 110°(230°F) Refractive Index: 1.5440 Boiling Point: 136-140°/16mm Beilstein Reference: 2045903 MDL No.: MFCD00001378

Formula: C₁₂H₁₄O
Boiling Pt: 136…140 °C/16 mm
Density: 1.036
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001378
CAS Number: 5422-88-8
EINECS: 226-548-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR