3-Cyanopyridine-N-oxide 97%

Supplier: Thermo Scientific Chemicals
Warning

A19961-09 A19961-88
CAAAA19961-09EA 183.28 CAD
CAAAA19961-09 CAAAA19961-88
3-Cyanopyridine-N-oxide 97%
3-Cyanopyridine N-oxide

Purity: 97% CAS No.: 14906-64-0 Molecular Formula: C6H4N2O Formula Weight: 120.11 Melting Point: 175-178° MDL No.: MFCD00129037

Formula: C₆H₄N₂O
Melting Pt: 175…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00129037
CAS Number: 14906-64-0
EINECS: 238-975-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR