2-Methyl-4-octanone 99%
Supplier: Thermo Scientific Chemicals
A19952-06
A19952-14
CAAAA19952-06EA
141.88
CAD
CAAAA19952-06
CAAAA19952-14
2-Methyl-4-octanone 99%
2-Methyl-4-octanone
Purity: 99%
CAS No.: 7492-38-8
Molecular Formula: C9H18O
Formula Weight: 142.24
Refractive Index: 1.4150
Boiling Point: 180°
MDL No.: MFCD00048808
Formula:
C₉H₁₈O Boiling Pt: 180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00048808 CAS Number: 7492-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...