4-Amino-2-(methylthio)pyrimidine-5-carbonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-5-cyano-2-(methylthio)pyrimidine

A19949-06 A19949-14
CAAAA19949-03EA 77.2 CAD
CAAAA19949-03 CAAAA19949-06 CAAAA19949-14
4-Amino-2-(methylthio)pyrimidine-5-carbonitrile 97%
4-Amino-2-(methylthio)pyrimidine-5-carbonitrile

Purity: 97% CAS No.: 770-30-9 Molecular Formula: C6H6N4S Formula Weight: 166.21 Melting Point: 240-243° UN No.: 3439 MDL No.: MFCD00023237

Formula: C₆H₆N₄S
MW: 166.21 g/mol
Melting Pt: 240…243 °C
Storage Temperature: Ambient
MDL Number: MFCD00023237
CAS Number: 770-30-9
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR