Brucine sulfate hydrate, (∼13% H₂O) 98% (dry weight)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 10,11-Dimethoxystrychnine sulphate salt, 10,11-Dimethoxystrychnine sulfate salt

A19943-14 A19943-22
CAAAAA19943-14EA 89.43 CAD
CAAAAA19943-14 CAAAAA19943-22
Brucine sulfate hydrate, (∼13% H₂O) 98% (dry weight)
Brucine sulfate hydrate

Purity: 98% CAS No.: 652154-10-4 Molecular Formula: C23H26N2O4.0.5H2SO4.xH2O Formula Weight: 443.52(anhy) Melting Point: 191-196° dec. UN No.: 2811 MDL No.: MFCD00013472

Formula: C₂₃H₂₆N₂O₄·¹/₂H₂SO₄·xH₂O
MW: 443.52 g/mol (anhy)
Melting Pt: 191…196 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150158
CAS Number: 652154-10-4
EINECS: 225-432-9
UN: 2811
ADR: "6.1",III
Merck Index: 14,1455

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR