Sodium-3-bromopropanesulfonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19930-06
A19930-14
A19930-22
CAAAA19930-06EA
70.31
CAD
CAAAA19930-06
CAAAA19930-14
CAAAA19930-22
Sodium-3-bromopropanesulfonate 97%
Sodium-3-bromopropanesulfonate
Purity: 97%
CAS No.: 55788-44-8
Molecular Formula: Br(CH2)3SO3Na
Formula Weight: 225.05
Melting Point: ca 256° dec.
MDL No.: MFCD00024999
Formula:
C₃H₇BrO₃S Melting Pt: ∼ 256 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00024999 CAS Number: 55788-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...