3,4-Dimethyl-1-pentyn-3-ol 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19906-06
A19906-14
CAAAA19906-06EA
61.62
CAD
CAAAA19906-06
CAAAA19906-14
3,4-Dimethyl-1-pentyn-3-ol 94%
3,4-Dimethyl-1-pentyn-3-ol
Purity: 94%
CAS No.: 1482-15-1
Molecular Formula: (CH3)2CHC(OH)(CH3)C≡CH
Formula Weight: 112.17
Flash Point: 40°(104°F)
Boiling Point: 133°
UN No.: 1987
MDL No.: MFCD00039845
Formula:
C₇H₁₂O Boiling Pt: 133 °C Density: 0.87 Flash Pt: 40 °C(104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039845 CAS Number: 1482-15-1 UN: 1987 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...