Isoquinoline-3-carboxylic acid monohydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19880-03
A19880-06
CAAAA19880-03EA
162.74
CAD
CAAAA19880-03
CAAAA19880-06
CAAAA19880-14
Isoquinoline-3-carboxylic acid monohydrate 99%
Isoquinoline-3-carboxylic acid monohydrate
Purity: 99%
CAS No.: 207399-25-5
Molecular Formula: C10H7NO2.xH2O
Formula Weight: 173.17(anhy)
Melting Point: 166-168°
Beilstein Reference: 125548
MDL No.: MFCD00167216
Formula:
C10H9NO3 Melting Pt: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167216 CAS Number: 207399-25-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...