Citronellyl acetate 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
3,7-Dimethyloct-6-en-1-yl acetate, 3,7-Dimethyl-6-octen-1-ol Acetate
A19872-14
A19872-22
CAAAA19872-14EA
67.94
CAD
CAAAA19872-14
CAAAA19872-22
Citronellyl acetate 96%
Citronellyl acetate
Purity: 96%
CAS No.: 150-84-5
Molecular Formula: C13H24O2
Formula Weight: 212.33
Flash Point: 98°(208°F)
Refractive Index: 1.4434
Boiling Point: 240°
MDL No.: MFCD00015039
Formula:
C₁₂H₂₂O₂ MW: 198.31 g/mol Boiling Pt: 240 °C Density: 0.890 Flash Pt: 98 °C(208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015039 CAS Number: 150-84-5 EINECS: 205-775-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...