5-Chloro-2-mercaptobenzimidazole 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Chloro-2-benzimidazolethiol

A19852-06 A19852-14
CAAAA19852-06EA 132.4 CAD
CAAAA19852-06 CAAAA19852-14
5-Chloro-2-mercaptobenzimidazole 98%
5-Chloro-2-mercaptobenzimidazole

Purity: 98% CAS No.: 25369-78-2 Molecular Formula: C7H5ClN2S Formula Weight: 184.64 Melting Point: 298-302° UN No.: 2811 MDL No.: MFCD01658762

Formula: C₇H₅ClN₂S
MW: 184.64 g/mol
Melting Pt: 298…302 °C
Storage Temperature: Ambient
MDL Number: MFCD01658762
CAS Number: 25369-78-2
EINECS: 246-903-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR