Methyl-5-methylisoxazole-3-carboxylate 97%
Supplier: Thermo Scientific Chemicals
A19814-14
A19814-22
CAAAA19814-14EA
186.44
CAD
CAAAA19814-14
CAAAA19814-22
CAAAA19814-36
Methyl-5-methylisoxazole-3-carboxylate 97%
Methyl 5-methylisoxazole-3-carboxylate
Purity: 97%
CAS No.: 19788-35-3
Molecular Formula: C6H7NO3
Formula Weight: 141.13
Melting Point: 91-92°
Boiling Point: 125°/11mm
MDL No.: MFCD00015895
Formula:
C₆H₇NO₃ Boiling Pt: 125 °C/11 mm Melting Pt: 91…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015895 CAS Number: 19788-35-3 EINECS: 243-311-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...