4,5-Dibromothiophene-2-carboxaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19788-06
A19788-14
CAAAA19788-06EA
189.6
CAD
CAAAA19788-06
CAAAA19788-14
4,5-Dibromothiophene-2-carboxaldehyde 98%
4,5-Dibromothiophene-2-carboxaldehyde
Purity: 98%
CAS No.: 38071-22-6
Molecular Formula: C5H2Br2OS
Formula Weight: 269.94
Melting Point: 79-81°
MDL No.: MFCD00225242
Formula:
C₅H₂Br₂OS Melting Pt: 79…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00225242 CAS Number: 38071-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...