Farnesyl acetate (mixture of isomers) mixture of isomers 96%
Supplier: Thermo Scientific Chemicals
A19778-14
A19778-22
CAAAA19778-14EA
121.5
CAD
CAAAA19778-14
CAAAA19778-22
Farnesyl acetate (mixture of isomers) mixture of isomers 96%
Farnesyl acetate (mixture of isomers)
Purity: 96%
CAS No.: 29548-30-9
Molecular Formula: C17H28O2
Formula Weight: 264.41
Flash Point: 95°(203°F)
Refractive Index: 1.477
Beilstein Reference: 1784823
MDL No.: MFCD00036516
Formula:
C₁₇H₂₈O₂ Density: 0.91 Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036516 CAS Number: 29548-30-9 EINECS: 249-689-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...